Learn More
Eurofins Discoverx PathHunter CHO-K1 SSTR2 β-Arrestin Cell Line, 2 vials. (93-0181C2)
Catalog No. 501999879
Encompass
The PathHunter SSTR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. somatostatin) based activation of SSTR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged SSTR2 and EA-tagged β-Arrestin-2. Activation of SSTR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9879 Supplier Eurofins Discoverx Supplier No. 930181C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter SSTR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. somatostatin) based activation of SSTR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged SSTR2 and EA-tagged β-Arrestin-2. Activation of SSTR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.