Learn More
Eurofins Discoverx PathHunter CHO-K1 TACR2 β-Arrestin Cell Line, 2 vials. (93-0238C2)
Catalog No. 502000043
Encompass
The PathHunter TACR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. substance P) based activation of TACR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TACR2 and EA-tagged β-Arrestin-2. Activation of TACR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0043 Supplier Eurofins Discoverx Supplier No. 930238C2
Add to Cart
Add to Cart
Request a Quote
The PathHunter TACR2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. substance P) based activation of TACR2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TACR2 and EA-tagged β-Arrestin-2. Activation of TACR2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.