Learn More
Eurofins Discoverx PathHunter CHO-K1 TRHR β-Arrestin Cell Line, 2 vials. (93-0239C2)
Supplier:  Eurofins Discoverx 930239C2
Encompass
The PathHunter TRHR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. TRH) based activation of TRHR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TRHR and EA-tagged β-Arrestin-2. Activation of TRHR stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0047
Add to Cart
Request a Quote