Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 TSHR (Long Isoform) Gs Cell Line, 2 vials. (95-0153C2)
Catalog No. 502003813
Encompass
The cAMP Hunter TSHRL Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. TSH) based activation of TSHRL GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type TSHRL, a naturally Gs coupled GPCR. Upon activation of TSHRL, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3813 Supplier Eurofins Discoverx Supplier No. 950153C2
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter TSHRL Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. TSH) based activation of TSHRL GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type TSHRL, a naturally Gs coupled GPCR. Upon activation of TSHRL, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.