Learn More
Eurofins Discoverx cAMP Hunter CHO-K1 VIPR1 Gs Cell Line, 2 vials. (95-0093C2)
Supplier: Eurofins Discoverx 950093C2
The cAMP Hunter VIPR1 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. VIP) based activation of VIPR1 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type VIPR1, a naturally Gs coupled GPCR. Upon activation of VIPR1, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.