Learn More
Crescent Chemical Co Inc CHOLAMIDOPROPYL)DIMETHYLAMMONI
SDP
Catalog No. 50247433
Encompass_Preferred
Cholamidopropyl)dimethylammonio]-1-propanesulfonate research grade Zwitterionic surfactant. Particularly useful for solubilizing and electrophoresis of membrane proteins (1 - 5). Suitable also for enzyme immunoassay (6). Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Numbe
Catalog No. 50-247-433 Supplier Crescent Chemical Co Inc Supplier No. SE17038.04
Cholamidopropyl)dimethylammonio]-1-propanesulfonate research grade Zwitterionic surfactant. Particularly useful for solubilizing and electrophoresis of membrane proteins (1 - 5). Suitable also for enzyme immunoassay (6). Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Numbe