Learn More
Crescent Chemical Co Inc CHOLAMIDOPROPYL)DIMETHYLAMMONI
SDP
Supplier:  Crescent Chemical Co Inc SE17038.03
Encompass_Preferred
Cholamidopropyl)dimethylammonio]-1-propanesulfonate research grade Zwitterionic surfactant. Particularly useful for solubilizing and electrophoresis of membrane proteins (1 - 5). Suitable also for enzyme immunoassay (6). Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number
Catalog No. 50-247-432