Learn More
Medchemexpress LLC Cholan-24-oic-2,2,4,4-d4 acid, 3-hydroxy-, (3α,5β)- | 83701-16-0 | 99.7% | 5 MG
SDP
Catalog No. 5000406675
Encompass_Preferred
Lithocholic acid-d4 is a deuterium labeled form of lithocholic acid, which is a toxic secondary bile acid. It appears as a white to off-white solid and is intended for research use only.
  • Deuterium labeled compound
  • Suitable for use as a tracer
  • Functions as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Presented as a white to off-white solid
Catalog No. 50-004-06675 Supplier Medchemexpress LLC Supplier No. HYB0172S5MG
Lithocholic acid-d4 is a deuterium labeled form of lithocholic acid, which is a toxic secondary bile acid. It appears as a white to off-white solid and is intended for research use only.
  • Deuterium labeled compound
  • Suitable for use as a tracer
  • Functions as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Presented as a white to off-white solid