Learn More
Medchemexpress LLC Cholan-24-oic-24-13C acid, 3,12-dihydroxy-, (3α,5β,12α)- | 52886-37-0 | MFCD00190301 | 98.0% | 393.56 | C24H40O4 | 10 MG
SDP
Catalog No. 5000298088
Encompass_Preferred
Deoxycholic acid-13C is the 13C-stable isotope labeled form of deoxycholic acid used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It also activates the G protein-coupled bile acid receptor TGR5 and is used in studies of bile acid signaling and brown adipose tissue activity.
  • 13C-stable isotope labeled deoxycholic acid for tracer studies and quantitation.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS methods.
  • Purity approximately 98.0% as supplied.
  • Available in small analytical quantities (for example, 10 mg).
  • Store powder at -20 °C for long-term stability; follow SDS for handling.
Catalog No. 50-002-98088 Supplier Medchemexpress LLC Supplier No. HYN0593S310MG
Deoxycholic acid-13C is the 13C-stable isotope labeled form of deoxycholic acid used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It also activates the G protein-coupled bile acid receptor TGR5 and is used in studies of bile acid signaling and brown adipose tissue activity.
  • 13C-stable isotope labeled deoxycholic acid for tracer studies and quantitation.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS methods.
  • Purity approximately 98.0% as supplied.
  • Available in small analytical quantities (for example, 10 mg).
  • Store powder at -20 °C for long-term stability; follow SDS for handling.