Learn More
Medchemexpress LLC Cholan-24-oic-24-13C acid, 3,7,12-trihydroxy-, (3α,5β,7α,12α)- | 52886-36-9 | 99.2% | 409.56 | C23(13C)H40O5 | 10 MG
SDP
Catalog No. 5000295841
Encompass_Preferred
Cholic acid-13C is the 13C-stable isotope labeled form of cholic acid, provided as a high-purity analytical standard for metabolic and lipid research. Supplied as a small quantity powder for laboratory use; not for human consumption.
  • 13C-stable isotope labeling for use in mass spectrometry and tracer studies.
  • High reported purity, suitable for analytical applications.
  • Supplied in a 10 mg quantity for standards and small-scale experiments.
  • Datasheet, certificate of analysis, and SDS available for quality and safety verification.
  • For research use only; not for human or therapeutic use.
Catalog No. 50-002-95841 Supplier Medchemexpress LLC Supplier No. HYN0324S210MG
Cholic acid-13C is the 13C-stable isotope labeled form of cholic acid, provided as a high-purity analytical standard for metabolic and lipid research. Supplied as a small quantity powder for laboratory use; not for human consumption.
  • 13C-stable isotope labeling for use in mass spectrometry and tracer studies.
  • High reported purity, suitable for analytical applications.
  • Supplied in a 10 mg quantity for standards and small-scale experiments.
  • Datasheet, certificate of analysis, and SDS available for quality and safety verification.
  • For research use only; not for human or therapeutic use.