Learn More
Cayman Chemical Cholic Acid, Purity: ≥95%, CAS: 81-25-4, 50 g
SDP
Catalog No. 502983591
Encompass_Preferred
A primary bile acid; induces C. difficile colony formation in an agar dilution assay at 0.1% w/v; dietary administration increases serum cholesterol levels, biliary phospholipid secretion, and fecal DCA levels in rats at 0.4% w/w
Catalog No. 50-298-3591 Supplier Cayman Chemical Supplier No. 2025050 G
A primary bile acid; induces C. difficile colony formation in an agar dilution assay at 0.1% w/v; dietary administration increases serum cholesterol levels, biliary phospholipid secretion, and fecal DCA levels in rats at 0.4% w/w