Learn More
Medchemexpress LLC Cholecalciferol | 67-97-0 | MFCD00078131 | 99.8% | 384.64 g/mol | C27H44O | 10 MG
SDP
Catalog No. 5000287413
Encompass_Preferred
Vitamin D3 (cholecalciferol) analytical standard supplied as a 10 mg vial for research and analytical applications. It is intended as a reference standard for assay calibration and method development in HPLC, GC, and MS; not for human consumption.
  • Analytical standard for assay calibration and method development.
  • High purity (99.8%).
  • Suitable for HPLC, GC, and mass spectrometry workflows.
  • Supplied in small pack sizes for laboratory use.
  • Chemical formula C27H44O; molecular weight 384.64 g/mol.
Catalog No. 50-002-87413 Supplier Medchemexpress LLC Supplier No. HY15398R10MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vitamin D3 (cholecalciferol) analytical standard supplied as a 10 mg vial for research and analytical applications. It is intended as a reference standard for assay calibration and method development in HPLC, GC, and MS; not for human consumption.
  • Analytical standard for assay calibration and method development.
  • High purity (99.8%).
  • Suitable for HPLC, GC, and mass spectrometry workflows.
  • Supplied in small pack sizes for laboratory use.
  • Chemical formula C27H44O; molecular weight 384.64 g/mol.