Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholecalciferol (D3), MilliporeSigma™ Supelco™

Catalog No. 111020535 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0535 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0535 Supplier MilliporeSigma Supelco Supplier No. 47763
Add to Cart
Add to Cart

Cholecalciferol (D3) is a fat soluble vitamin, and is one of the forms of vitamin D, synthesized in human skin on exposure to ultraviolet (UV) radiation from sunlight, or it can also be obtained from food. It is found to be essential in humans for maintaining normal calcium metabolism.

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 67-97-0
Physical Form Neat
Linear Formula C27H44 O
MDL Number MFCD00078131
Melting Point 83°C to 86°C
Synonym Cholecalciferol; (+)-Vitamin D3; 7-Dehydrocholesterol activated; Activated 7-dehydrocholesterol; Calciol
Type Food and Cosmetic Component Standard
Quantity 100 mg
Formula Weight 384.64
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.