Learn More
Abcam Cholecalciferol (Vitamin D3), VDR activator, 5G
Catalog No. 502706106
Encompass_Preferred
MW 384.6 Da, Purity >98%. VDR activator. Induces cell differentiation and shows antiproliferative effects. Metabolic calcifediol (ab142766) precursor. Shows antioxidant and anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6106 Supplier Abcam Supplier No. AB1435945G
MW 384.6 Da, Purity >98%. VDR activator. Induces cell differentiation and shows antiproliferative effects. Metabolic calcifediol (ab142766) precursor. Shows antioxidant and anti-inflammatory effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement