Learn More
Medchemexpress LLC Cholecystokinin (26-33) free acid | 103974-46-5 | 99.8% | 1064.19 | 1 MG

Supplier: Medchemexpress LLC HYP36511MG
Cholecystokinin (26-33) (CCK (26-33)) free acid is a cholecystokinin (CCK) fragment. It is known to reduce food intake and stimulate gallbladder contraction.
- Purity of 99.8%
- Molecular weight of 1064.19
- Available as an off-white to pink solid
- Store powder at -80°C for 2 years or -20°C for 1 year
- Store in solvent at -80°C for 6 months or -20°C for 1 month
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.