Learn More
CPC Scientific H-Tyr-Met-Gly-Trp-Met-Asp-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CCKS007B
Encompass
Heptapeptide.ONE-LETTER SEQUENCE: YMGWMDF-NH2MOLECULAR FORMULA: C45H57N9O10S2MOLECULAR WEIGHT:948.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [47910-79-2], RESEARCH AREA: GastrointestinalREFERENCES: P.W. Schiller et al., BBRC, 85, 1332 (1978)
Catalog No. 50-210-9122