Learn More
CPC Scientific H-Trp-Met-Asp-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CCKS008B
Encompass
Potent releaser of insulin and other islet hormones. ONE-LETTER SEQUENCE: WMDF-NH2MOLECULAR FORMULA: C29H36N6O6S1MOLECULAR WEIGHT:596.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [5609-49-4], RESEARCH AREA: GastrointestinalREFERENCES: J.F. Rehfeld et al., Nature, 284, 33 (1980)
Catalog No. 50-210-9124