Learn More
CPC Scientific H-Ser-Ala-Glu-Glu-Tyr-Glu-Tyr-Pro-Ser-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CCKS009B
Encompass
ONE-LETTER SEQUENCE: SAEEYEYPSMOLECULAR FORMULA: C47H63N9O20MOLECULAR WEIGHT:1074.05STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [198483-37-3], RESEARCH AREA: Gastrointestinal
Catalog No. 50-210-9126