Learn More
Cayman Chemical CholenIc AcId 100mg
SDP
Catalog No. 502991971
Encompass_Preferred
A bile acid; formed by 7α-hydroxylation of 27-hydroxycholesterol; precursor in the biosynthesis of chenodeoxycholic acid; levels are increased in patients with neonatal liver disease harboring mutations in CYP7A1, as well as in patients with intrahepatic and extrahepatic cholestasis
Catalog No. 50-299-1971 Supplier Cayman Chemical Supplier No. 29543100 MG
A bile acid; formed by 7α-hydroxylation of 27-hydroxycholesterol; precursor in the biosynthesis of chenodeoxycholic acid; levels are increased in patients with neonatal liver disease harboring mutations in CYP7A1, as well as in patients with intrahepatic and extrahepatic cholestasis