Learn More
Cayman Chemical Cholenic Acid, Purity: ≥98%, CAS: 5255-17-4, 500 mg
SDP
Catalog No. 502991149
Encompass_Preferred
A bile acid; formed by 7α-hydroxylation of 27-hydroxycholesterol; precursor in the biosynthesis of chenodeoxycholic acid; levels are increased in patients with neonatal liver disease harboring mutations in CYP7A1, as well as in patients with intrahepatic and extrahepatic cholestasis
Catalog No. 50-299-1149 Supplier Cayman Chemical Supplier No. 29543500 MG
A bile acid; formed by 7α-hydroxylation of 27-hydroxycholesterol; precursor in the biosynthesis of chenodeoxycholic acid; levels are increased in patients with neonatal liver disease harboring mutations in CYP7A1, as well as in patients with intrahepatic and extrahepatic cholestasis