Learn More
Bioss Cholera toxin
SDP
Catalog No. 5000065859
Encompass_Preferred
Cholera toxin Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65859 Supplier Bioss Supplier No. BSM72061M
Cholera toxin Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide