Learn More
Biotium Cholera Toxin B, CF594 Conjugate, 100 ug
Catalog No. 501964085
Encompass_Preferred
Cholera toxin B subunit is used for tract-tracing in neurological research, targeting GM1 ganglioside binding and retrograde transport. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Deep-red fluorescent CF594 dye has excitation/emission at 593/614 nm.
Catalog No. 50-196-4085 Supplier Biotium Supplier No. 00072
Cholera toxin B subunit is used for tract-tracing in neurological research, targeting GM1 ganglioside binding and retrograde transport. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Deep-red fluorescent CF594 dye has excitation/emission at 593/614 nm.