Learn More
Sigma Aldrich Fine Chemicals Biosciences Cholera Toxin B subunit 955MG
Catalog No. 502891550
Encompass
Cholera Toxin is a heterohexameric AB5 enterotoxin. Cholera Toxin B subunit is the the B subunit pentamer of cholera toxin produced by Vibrio cholera. It is generally involved in the cell trafficking and signalling machineries.
Catalog No. 50-289-1550 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C9903.5MG
 Restricted Item
Add to Cart
Add to Cart
Cholera Toxin is a heterohexameric AB5 enterotoxin. Cholera Toxin B subunit is the the B subunit pentamer of cholera toxin produced by Vibrio cholera. It is generally involved in the cell trafficking and signalling machineries.