Learn More
Sigma Aldrich Fine Chemicals Biosciences Cholera Toxin B subunit bi5MG
Catalog No. 502905299
Encompass
Cholera Toxin B (CTB) is secreted by Vibrio cholerae. CTB functions as an oral subunit vaccine for cholera which is associated with acute watery diarrhoea. It acts as a mucosal immunogen. Cholera toxin (CT) stimulates cell surface molecules such as antigen presenting cells (APCs) murine and human dendritic cells (DCs). CT also has immunomodulatory properties. It induces the secretion of interleukin 1 (IL-1) from macrophages and enhances their APC function.
Catalog No. 50-290-5299 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C9972.5MG
Cholera Toxin B (CTB) is secreted by Vibrio cholerae. CTB functions as an oral subunit vaccine for cholera which is associated with acute watery diarrhoea. It acts as a mucosal immunogen. Cholera toxin (CT) stimulates cell surface molecules such as antigen presenting cells (APCs) murine and human dendritic cells (DCs). CT also has immunomodulatory properties. It induces the secretion of interleukin 1 (IL-1) from macrophages and enhances their APC function.