Learn More
Sigma Aldrich Fine Chemicals Biosciences Cholera Toxin B subunit re
Catalog No. 502905309
Encompass
The advantage of this product over the native CTXB is that it has no traces of Vibrio cholera and no contamination of the highly toxic Cholera toxin A subunit. This product is expressed in human HEK 293 cells as a C-terminally his-tagged protein with a calculated molecular mass of 13 kDa (amino acids Thr22-Asn124).
Catalog No. 50-290-5309 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE0069500UG
The advantage of this product over the native CTXB is that it has no traces of Vibrio cholera and no contamination of the highly toxic Cholera toxin A subunit. This product is expressed in human HEK 293 cells as a C-terminally his-tagged protein with a calculated molecular mass of 13 kDa (amino acids Thr22-Asn124).