Learn More
Medchemexpress LLC Cholera toxin B subunit, from vibrio cholerae | 131096-89-4 | 95.0% | 1 MG
SDP
Catalog No. 5000396166
Encompass_Preferred
Cholera toxin B subunit, from vibrio cholerae (CTB) is non-toxic to cells and lacks intrinsic adenylate cyclase activity. It attaches to cells by binding to ganglioside GM1. CTB is recognized as an effective tracer for studying axonal transport and is widely used as a marker for membrane lipid rafts.
  • Attaches to cells by binding to ganglioside GM1
  • Suitable label for microglial cells
  • Effective tracer for studying axonal transport using immunohistochemical methods
  • Widely used as a marker of membrane lipid rafts
  • Inhibits the production of TNFα in E. coli
Catalog No. 50-003-96166 Supplier Medchemexpress LLC Supplier No. HYP33941MG
Cholera toxin B subunit, from vibrio cholerae (CTB) is non-toxic to cells and lacks intrinsic adenylate cyclase activity. It attaches to cells by binding to ganglioside GM1. CTB is recognized as an effective tracer for studying axonal transport and is widely used as a marker for membrane lipid rafts.
  • Attaches to cells by binding to ganglioside GM1
  • Suitable label for microglial cells
  • Effective tracer for studying axonal transport using immunohistochemical methods
  • Widely used as a marker of membrane lipid rafts
  • Inhibits the production of TNFα in E. coli