Learn More
Cayman Chemical Cholera Toxin (azide free), Purity: ≥95% (estimated by SDS-PAGE), 1 mg
SDP
Catalog No. 502984225
Encompass_Preferred
A multimeric enterotoxin that catalyzes the ADP-ribosylation of the Gsα protein, which activates adenylate cyclase and increases the intracellular concentration of cAMP; causes the hypersecretion of chloride and bicarbonate ions, which results in increased fluid secretion and the severe diarrhea characteristic of the disease, cholera
Catalog No. 50-298-4225 Supplier Cayman Chemical Supplier No. 196541 MG
A multimeric enterotoxin that catalyzes the ADP-ribosylation of the Gsα protein, which activates adenylate cyclase and increases the intracellular concentration of cAMP; causes the hypersecretion of chloride and bicarbonate ions, which results in increased fluid secretion and the severe diarrhea characteristic of the disease, cholera