Learn More
American Radiolabeled Chemicals Inc CHOLES STEAR 3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 194050UC
Encompass_Preferred
Cholesteryl stearate-cholesterol-1,2-3H 50uCi in toluene in sealed ampoule, 1 mCi/ml, 50-60 Ci/mmol, M.W. 653.12, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-920