Learn More
Medchemexpress LLC Cholest-5-en-3-ol (3ẞ)-, 3-[12-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]dodecanoate] | 1246303-05-8 | 99.9% | 747.06 | 500 UG
SDP
Catalog No. 5000352024
Encompass_Preferred
3-C12-NBD-cholesterol is a fluorescent cholesterol analog designed for measuring membrane and intracellular trafficking dynamics.
  • Fluorescent cholesterol analog
  • Used for measuring membrane and intracellular trafficking dynamics
  • Appearance: colorless to light yellow liquid
  • High purity (99.9%)
Catalog No. 50-003-52024 Supplier Medchemexpress LLC Supplier No. HY139407500UG
3-C12-NBD-cholesterol is a fluorescent cholesterol analog designed for measuring membrane and intracellular trafficking dynamics.
  • Fluorescent cholesterol analog
  • Used for measuring membrane and intracellular trafficking dynamics
  • Appearance: colorless to light yellow liquid
  • High purity (99.9%)