Learn More
Medchemexpress LLC Cholest-5-en-3-ol (3β)-3-[6-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]hexanoate] | 201731-19-3 | MFCD18428012 | 99.0% | 662.90 g/mol | C39H58N4O5 | 5 MG
SDP
Catalog No. 5000263055
Encompass_Preferred
3-C6-NBD-cholesterol is a fluorescently labeled cholesterol analogue used to monitor membrane insertion and intracellular cholesterol trafficking in fluorescence-based assays. The NBD fluorophore attached via a six-carbon linker provides environment-sensitive emission useful for kinetic and localization studies.
  • Fluorescent cholesterol analogue for monitoring membrane insertion and intracellular trafficking.
  • NBD fluorophore attached via a six-carbon linker for environment-sensitive fluorescence.
  • High purity (99.0%) for reproducible experimental results.
  • Solid, yellow to orange appearance suitable for handling and formulation.
  • Stable as powder at -20°C for long-term storage.
  • Provided in small research packs for assay development and optimization.
Catalog No. 50-002-63055 Supplier Medchemexpress LLC Supplier No. HY1316825MG
3-C6-NBD-cholesterol is a fluorescently labeled cholesterol analogue used to monitor membrane insertion and intracellular cholesterol trafficking in fluorescence-based assays. The NBD fluorophore attached via a six-carbon linker provides environment-sensitive emission useful for kinetic and localization studies.
  • Fluorescent cholesterol analogue for monitoring membrane insertion and intracellular trafficking.
  • NBD fluorophore attached via a six-carbon linker for environment-sensitive fluorescence.
  • High purity (99.0%) for reproducible experimental results.
  • Solid, yellow to orange appearance suitable for handling and formulation.
  • Stable as powder at -20°C for long-term storage.
  • Provided in small research packs for assay development and optimization.