Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cholest-5-ene-3ß,22(S)-diol | 22348-64-7 | 98.6% | 402.7 g/mol | C27H46O2 | 5 MG
SDP

Catalog No. 5000221977
Encompass_Preferred

Cholest-5-ene-3ß,22(S)-diol ((22S)-hydroxycholesterol) is an oxysterol research reagent used in studies of cholesterol metabolism and lipid biology. Manufacturer data report it as orally active, with no significant cytotoxic, oxidative, or inflammatory effects observed in tested cell models, and with effects on lipid endpoints in rodent studies.

  • Purity 98.6%.
  • Chemical formula C27H46O2.
  • Molecular weight 402.7 g/mol.
  • Available as a 5 mg vial.
  • Orally active oxysterol with reported lipid-modulating activity in rodent models.
  • Minimal cytotoxic, oxidative, or inflammatory effects reported in tested cell models.
  • Suitable for studies of cholesterol metabolism and lipid signaling pathways.

Catalog No. 50-002-21977 Supplier Medchemexpress LLC Supplier No. HY1373075MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cholest-5-ene-3ß,22(S)-diol ((22S)-hydroxycholesterol) is an oxysterol research reagent used in studies of cholesterol metabolism and lipid biology. Manufacturer data report it as orally active, with no significant cytotoxic, oxidative, or inflammatory effects observed in tested cell models, and with effects on lipid endpoints in rodent studies.

  • Purity 98.6%.
  • Chemical formula C27H46O2.
  • Molecular weight 402.7 g/mol.
  • Available as a 5 mg vial.
  • Orally active oxysterol with reported lipid-modulating activity in rodent models.
  • Minimal cytotoxic, oxidative, or inflammatory effects reported in tested cell models.
  • Suitable for studies of cholesterol metabolism and lipid signaling pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.