Learn More
Cayman Chemical Cholesta-3,5-dien-7-one, Purity: ≥95%, CAS: 567-72-6, 10 mg
SDP
Catalog No. 502946273
Encompass_Preferred
An oxysterol and a negative allosteric modulator of GABAA receptors; reduces GABA-induced currents in HEK cells expressing α1β1γ2 or α4β3γ2 subunit-containing GABAA receptors (IC50s = 1.5 and 1 µM, respectively); reduces GABA-induced depolarization of peptidergic and non-peptidergic nociceptors, C-LTMRs, and cold thermosensors in isolated mouse DRG neurons; increases latency to nocifensive behaviors in the hot plate test in mice at 2, 10, and 50 mg/kg
Catalog No. 50-294-6273 Supplier Cayman Chemical Supplier No. 3432010 MG
An oxysterol and a negative allosteric modulator of GABAA receptors; reduces GABA-induced currents in HEK cells expressing α1β1γ2 or α4β3γ2 subunit-containing GABAA receptors (IC50s = 1.5 and 1 µM, respectively); reduces GABA-induced depolarization of peptidergic and non-peptidergic nociceptors, C-LTMRs, and cold thermosensors in isolated mouse DRG neurons; increases latency to nocifensive behaviors in the hot plate test in mice at 2, 10, and 50 mg/kg