Learn More
Medchemexpress LLC Cholesta-3,5-diene | 747-90-0 | MFCD00009939 | 99.5% | 368.64 g/mol | C27H44 | 10 MG
SDP
Catalog No. 5000283589
Encompass_Preferred
Cholesta-3,5-diene is an oxysterol research compound that modulates inflammation by promoting neutrophil chemotaxis and fibroblast migration, accelerating wound repair through chemokine receptor-mediated pathways such as PI3K/Akt. Supplied as a solid powder for laboratory use, it is intended for research applications investigating inflammatory responses and wound-healing mechanisms and is not for human therapeutic use.
  • Modulates inflammation and accelerates wound repair.
  • Promotes neutrophil chemotaxis and fibroblast migration.
  • Activates chemokine receptor-mediated PI3K/Akt signaling.
  • High purity, ~99.5% (manufacturer reported).
  • Solid powder formulation suitable for topical and in vitro assays.
  • CAS number 747-90-0; formula C27H44; MW 368.64 g/mol.
Catalog No. 50-002-83589 Supplier Medchemexpress LLC Supplier No. HYW12757510MG
Cholesta-3,5-diene is an oxysterol research compound that modulates inflammation by promoting neutrophil chemotaxis and fibroblast migration, accelerating wound repair through chemokine receptor-mediated pathways such as PI3K/Akt. Supplied as a solid powder for laboratory use, it is intended for research applications investigating inflammatory responses and wound-healing mechanisms and is not for human therapeutic use.
  • Modulates inflammation and accelerates wound repair.
  • Promotes neutrophil chemotaxis and fibroblast migration.
  • Activates chemokine receptor-mediated PI3K/Akt signaling.
  • High purity, ~99.5% (manufacturer reported).
  • Solid powder formulation suitable for topical and in vitro assays.
  • CAS number 747-90-0; formula C27H44; MW 368.64 g/mol.