Learn More
Medchemexpress LLC Cholesta-3,5-diene | 747-90-0 | MFCD00009939 | 99.5% | 368.64 g/mol | C27H44 | 5 MG
SDP
Catalog No. 5000282645
Encompass_Preferred
Cholesta-3,5-diene is a cholesterol-derived oxysterol used as a research reagent to modulate inflammatory responses. It influences immune cell behavior, notably neutrophil chemotaxis, and is applied in studies of wound healing and immune signaling. Supplied as a white to off-white solid with high purity (≈99.5%), it is intended for laboratory research use only.
  • High purity suitable for analytical and biological assays.
  • Modulates neutrophil chemotaxis and immune responses.
  • Solid form facilitates accurate weighing and solution preparation.
  • Stable when stored as a powder at -20°C for long-term use.
  • Available in small quantities for screening and pilot studies.
Catalog No. 50-002-82645 Supplier Medchemexpress LLC Supplier No. HYW1275755MG
Cholesta-3,5-diene is a cholesterol-derived oxysterol used as a research reagent to modulate inflammatory responses. It influences immune cell behavior, notably neutrophil chemotaxis, and is applied in studies of wound healing and immune signaling. Supplied as a white to off-white solid with high purity (≈99.5%), it is intended for laboratory research use only.
  • High purity suitable for analytical and biological assays.
  • Modulates neutrophil chemotaxis and immune responses.
  • Solid form facilitates accurate weighing and solution preparation.
  • Stable when stored as a powder at -20°C for long-term use.
  • Available in small quantities for screening and pilot studies.