Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cholesta-5,24-dien-26,26,26,27,27,27-d6-3-ol, (3β)- | 1246298-67-8 | 99.4% | 390.67 | 1 MG
SDP

Catalog No. 5000436115
Encompass_Preferred

Desmosterol-d6 is a deuterium-labeled version of Desmosterol, an endogenous metabolite that serves as the immediate precursor to cholesterol in the Bloch pathway of cholesterol biosynthesis. It is used to study cholesterol metabolism. This molecule has a molecular weight of 390.67 and a chemical formula of C27H38D6O. Its CAS number is 1246298-67-8.

  • Purity of 99.38%.
  • Can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Appears as a solid, white to off-white.
  • Soluble in DMF at concentrations of 50 mg/mL (127.99 mM) or greater.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only and not for human use.

Catalog No. 50-004-36115 Supplier Medchemexpress LLC Supplier No. HY113224S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Desmosterol-d6 is a deuterium-labeled version of Desmosterol, an endogenous metabolite that serves as the immediate precursor to cholesterol in the Bloch pathway of cholesterol biosynthesis. It is used to study cholesterol metabolism. This molecule has a molecular weight of 390.67 and a chemical formula of C27H38D6O. Its CAS number is 1246298-67-8.

  • Purity of 99.38%.
  • Can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Appears as a solid, white to off-white.
  • Soluble in DMF at concentrations of 50 mg/mL (127.99 mM) or greater.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only and not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.