Learn More
Medchemexpress LLC Cholesta-5,24-dien-26,26,26,27,27,27-d6-3-ol, (3β)- | 1246298-67-8 | 99.4% | 390.67 | 500 UG
SDP
Catalog No. 5000436221
Encompass_Preferred
Desmosterol-d6 is the deuterium-labeled form of Desmosterol. Desmosterol is a molecule similar to cholesterol and is the immediate precursor of cholesterol in the Bloch pathway of cholesterol biosynthesis. As an endogenous metabolite, Desmosterol-d6 is used to study cholesterol metabolism.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-36221 Supplier Medchemexpress LLC Supplier No. HY113224S500UG
Desmosterol-d6 is the deuterium-labeled form of Desmosterol. Desmosterol is a molecule similar to cholesterol and is the immediate precursor of cholesterol in the Bloch pathway of cholesterol biosynthesis. As an endogenous metabolite, Desmosterol-d6 is used to study cholesterol metabolism.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.