Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-cholestane-d4 | 205529-74-4 | MFCD00216162 | 99.0% | 376.69 g/mol | C27H44D4 | 5 MG
SDP

Catalog No. 5000262077
Encompass_Preferred

α-Cholestane-d4 is a deuterium-labeled analogue of α-cholestane supplied as a white to off-white solid for research use. It functions as a tracer and internal standard in analytical workflows such as NMR, GC-MS, and LC-MS, supporting quantitative measurements and metabolic studies.

  • Deuterium-labeled internal standard for tracer and quantitative analyses.
  • High purity (99.0%) suitable for analytical applications.
  • Molecular formula C27H44D4; molecular weight 376.69 g/mol.
  • Appearance: white to off-white solid.
  • Available in small-scale quantities (1 mg, 5 mg, 10 mg).
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C for extended storage.

Catalog No. 50-002-62077 Supplier Medchemexpress LLC Supplier No. HY132649S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Cholestane-d4 is a deuterium-labeled analogue of α-cholestane supplied as a white to off-white solid for research use. It functions as a tracer and internal standard in analytical workflows such as NMR, GC-MS, and LC-MS, supporting quantitative measurements and metabolic studies.

  • Deuterium-labeled internal standard for tracer and quantitative analyses.
  • High purity (99.0%) suitable for analytical applications.
  • Molecular formula C27H44D4; molecular weight 376.69 g/mol.
  • Appearance: white to off-white solid.
  • Available in small-scale quantities (1 mg, 5 mg, 10 mg).
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.