Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Capillary Plungers for Cholestech LDX™ System
GSA_VA

Catalog No. 23762976 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
For Use With (Equipment):
Cholestech LDX™ Lipid Monitoring System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-762-976 Cholestech LDX™ Lipid Monitoring System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-762-976 Supplier Abbott™ Supplier No. 10311
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

50 Capillary plungers

Accessory for use with Cholestech LDX™ System.

Accessory for use with Cholestech LDX lipid monitoring system.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Capillary Plunger
For Use With (Equipment) Cholestech LDX™ Lipid Monitoring System
Includes 50 Capillary plungers
Content And Storage 15°C to 30°C (59°F to 86°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.