Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Capillary Tube for Cholestech LDX™ System

Catalog No. 23763105 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Capacity (Metric):
40 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
23-763-105 40 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-763-105 Supplier Abbott™ Supplier No. 52193
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Lipids/Glucose/ALT/AST

  • Cholestech LDX Lipid Monitoring System
  • 50/Pk.

Specifications

Type Capillary Tubes, Lipids/ALT/AST
Sample Type Blood
For Use With (Equipment) All lipid, total Cholesterol and HS CRP test cassettes of Cholestech LDX™ analyzer
Capacity (Metric) 40 μL
Includes 50 Capillary tubes
Content And Storage 15°C to 30°C (59°F to 86°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.