Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Cassette Cartridges for Cholestech LDX™ System

Catalog No. 23762961 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Detects:
Lipid Profile/Glucose
Total Cholesterol/Glucose
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detects
23-762-961 Lipid Profile/Glucose
23-762-957 Total Cholesterol/Glucose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-762-961 Supplier Abbott™ Supplier No. 10991

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cassette cartridges for use with Cholestech LDX™ System.

  • Rapid: Yields results in 5 minutes
  • Flexible: fingerstick or venous whole blood
  • Convenient: Sample size is 40μL
  • CLIA-waived (for whole blood)

Specifications

Type Test Cassette
Sample Type Blood
For Use With (Equipment) Cholestech LDX™ Cholesterol analyzer
Analysis Time 5 min.
Detects Lipid Profile/Glucose
Capacity (Metric) 40 μL
Certifications/Compliance CLIA waived/professional use only
Includes 10 Individually wrapped test cassettes and package insert
Content And Storage 2°C to 8°C (36°F to 46°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.