Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Cholestech LDX™ Analyzer

Catalog No. 23762966 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Display:
LCD
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display
23-762-966 LCD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-762-966 Supplier Abbott™ Supplier No. 10959
Add to Cart
Add to Cart

The CLIA-waived Cholestech LDX™ Analyzer is engineered for confidence, providing accurate, actionable, and readily accessible results that have set the standard in point-of-care lipid profile, cholesterol, and glucose testing.

  • Perform lipid, glucose testing while the patient is still in the office
  • Same level of accuracy and precision provided by laboratories with a turnaround time of minutes instead of days
  • CRMLN certified
  • Three simple steps to perform a complete lipid profile
    1. Collect fingerstick sample in provided capillary tube
    2. Apply sample to disposable test cassette
    3. Press Run button — results are shown in five minutes

Specifications

Type LDX Lipid Monitoring System
Sample Type Capillary and Venous Blood
Analysis Time 5 min.
Detects Lipid Profile, Lipid Profile-Glucose
Calibration Yes
Certifications/Compliance CRMLN; LSP; CLIA waived/professional use only
Includes Analyzer, printer, power supply, starter pack (200 capillary tubes, 200 capillary plungers, one 40μL Mini-Pet pipette, 50 pipette tips, optics check cassette, printer cable, and training video.
Content And Storage 15°C to 30°C (59°F to 86°F)
Amperage 1 A
Voltage 9 VDC
Display LCD
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.