Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott Cholestech LDX TC/HDL/GLU Test Cassette

Catalog No. 23763181 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
10/Cs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-763-181 10/Cs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-763-181 Supplier Abbott Supplier No. 97990
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Cholestech LDX™ TC/HDL/GLU test is a CLIA-waived, point-of-care test for measuring total and HDL cholesterol plus glucose from a small 40 μL fingerstick whole blood sample. CRMLN and LSP Certified.

  • CLIA-Waived
  • Results in 5 minutes
  • Small fingerstick sample
  • For use with the Cholestech LDX™ System.
  • The Cholestech LDX TC/HDL/GLU Cassette measures total cholesterol (TC), a measure of the total amount of cholesterol in the blood, and high density lipoprotein (HDL) cholesterol (referred to as the 'good cholesterol') as well as glucose (GLU), a measure of blood sugar.
  • TC/HDL/GLU also calculates the TC/HDL ratio and non-HDL cholesterol.

Specifications

Calibration Using the built-in magnetic stripe
CPT Code 82465QW, 83718QW, 82947QW
CE Marker Yes
Clia Complexity Waived
Detectable Analytes Cholesterol
Format Cassette
Sample Type Whole Blood
Type TC/HDL/GLU
Disposable Single-use
Packaging Type Box
Quantity 10/Cs
Sample Size 40 μL
Test Time 5 minutes
Analysis Time 5 minutes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.