Learn More
Abbott™ MiniPet™ Pipette Tips
Catalog No. 23762979 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Disposable:
Single-use
Catalog No. Disposable
23-762-979 Single-use
1 options
Catalog No. 23-762-979 Supplier Abbott™ Supplier No. 11010
Add to Cart
Add to Cart
For use with MiniPet Pipettes.
  • Disposable
  • For use with 35 μL, 40 μL and 50 μL
Specifications
Disposable Single-use
For Use With (Equipment) Cholestech LDX System, MiniPet Disposable
Product Line MiniPet