Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alere Single-use Lancet for Cholesterol Analyzer

Catalog No. 23763400 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
For Use With (Equipment):
Cholestech LDX™ Lipid Monitoring System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-763-400 Cholestech LDX™ Lipid Monitoring System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-763-400 Supplier Alere Supplier No. 13791

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Cholestech LDX™ lipid monitoring system

  • Used to poke finger to draw blood
  • Single use
  • 200/Pk.

Specifications

Type Lancet
Sample Type Blood
For Use With (Equipment) Cholestech LDX™ Lipid Monitoring System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.