Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Test Cassette for Cholestech LDX™ System
GSA_VA

Catalog No. 23762964 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
For Use With (Equipment):
Cholestech LDX™ Cholesterol analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-762-964 Cholestech LDX™ Cholesterol analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-762-964 Supplier Abbott™ Supplier No. 10228
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

LDX Optics check cassette

Features Optics Check Cassette with Optics Storage Case

  • Temperature range: 15° to 30°C (59° to 86°F)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Test Cassette
For Use With (Equipment) Cholestech LDX™ Cholesterol analyzer
Capacity (Metric) 40 μL
Includes LDX Optics check cassette
Content And Storage 15°C to 30°C (59°F to 86°F)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.