Learn More
Medchemexpress LLC Cholestenone-13C | 68165-57-1 | 99.0% | 385.63 | 1 MG
SDP
Catalog No. 5000436076
Encompass_Preferred
Cholestenone-13C is the 13C labeled Cholestenone. Cholestenone (4-Cholesten-3-one), the intermediate oxidation product of cholesterol, is metabolized primarily in the liver. Cholestenone is highly mobile in membranes and influences cholesterol flip-flop and efflux. Cholestenone may cause long-term functional defects in cells. It is for research use only and not sold to patients.
  • 13C labeled Cholestenone.
  • Intermediate oxidation product of cholesterol.
  • Highly mobile in membranes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration (incorporation of stable heavy isotopes) has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-36076 Supplier Medchemexpress LLC Supplier No. HY113365S11MG
Cholestenone-13C is the 13C labeled Cholestenone. Cholestenone (4-Cholesten-3-one), the intermediate oxidation product of cholesterol, is metabolized primarily in the liver. Cholestenone is highly mobile in membranes and influences cholesterol flip-flop and efflux. Cholestenone may cause long-term functional defects in cells. It is for research use only and not sold to patients.
  • 13C labeled Cholestenone.
  • Intermediate oxidation product of cholesterol.
  • Highly mobile in membranes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration (incorporation of stable heavy isotopes) has potential to affect pharmacokinetic and metabolic profiles of drugs.