Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cholestenone-d5 (4-cholesten-3-one-d5) | 72560-60-2 | MFCD31814277 | 99.4% | 389.67 g/mol | C27H39D5O | 5 MG
SDP

Catalog No. 5000264151
Encompass_Preferred

Cholestenone-d5 (4-cholesten-3-one-d5) is a deuterated stable-isotope analogue of cholestenone used as an internal standard and tracer in mass spectrometry and lipid metabolism studies. Supplied as a purified powder in small pre-weighed quantities for analytical workflows, it is characterized for defined purity, solubility, and storage stability.

  • Deuterated internal standard for mass spectrometry.
  • Molecular formula C27H39D5O.
  • Molecular weight 389.67 g/mol.
  • Purity ~99.4% (99.42% documented).
  • Soluble in ethanol at 50 mg/mL (requires ultrasonic).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Available in small sizes including 5 mg.

Catalog No. 50-002-64151 Supplier Medchemexpress LLC Supplier No. HY113365S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cholestenone-d5 (4-cholesten-3-one-d5) is a deuterated stable-isotope analogue of cholestenone used as an internal standard and tracer in mass spectrometry and lipid metabolism studies. Supplied as a purified powder in small pre-weighed quantities for analytical workflows, it is characterized for defined purity, solubility, and storage stability.

  • Deuterated internal standard for mass spectrometry.
  • Molecular formula C27H39D5O.
  • Molecular weight 389.67 g/mol.
  • Purity ~99.4% (99.42% documented).
  • Soluble in ethanol at 50 mg/mL (requires ultrasonic).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Available in small sizes including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.