Learn More
Cayman Chemical Cholesterol, Purity: ≥95%, CAS: 57-88-5, 100 g
SDP
Catalog No. 502897026
Encompass_Preferred
A sterol; a component of mammalian cell membranes that interacts with membrane phospholipids, sphingolipids, and proteins; a precursor of steroid hormones, bile acids, and the active form of vitamin D
Catalog No. 50-289-7026 Supplier Cayman Chemical Supplier No. 9003100100 G
A sterol; a component of mammalian cell membranes that interacts with membrane phospholipids, sphingolipids, and proteins; a precursor of steroid hormones, bile acids, and the active form of vitamin D