Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholesterol 2,4-Dichlorobenzoate, TCI America™
SDP

Catalog No. C126025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C126025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C126025G Supplier TCI America Supplier No. C126025G

Chemical Identifiers

CAS 32832-01-2
Molecular Formula C34H48Cl2O2
Molecular Weight (g/mol) 559.656
MDL Number MFCD00065268
InChI Key NZZFKZMKJPWVDL-LPVBUYOKSA-N
Synonym cholesteryl-2,4-dichlor-benzoat, cholest-5-en-3??-yl 2,4-dichlorobenzoate, 3??-cholest-5-en-3-ol, 2,4-dichlorobenzoate
PubChem CID 122130190
IUPAC Name [(3S,9S,14S,17R)-10,13-dimethyl-17-(6-methylheptan-2-yl)-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] 2,4-dichlorobenzoate
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C5=C(C=C(C=C5)Cl)Cl)C)C

Specifications

Melting Point 135°C
Color White
Quantity 25 g
Formula Weight 559.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cholesterol 2,4-Dichlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.