Learn More
American Radiolabeled Chemicals Inc CHOLESTEROL 24 25 3H 1LC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 19871MC
Encompass_Preferred
Cholesterol-24,25-3H 1mCi in ethanol, 1 mCi/ml, 40-60 Ci/mmol, M.W. 386.6, Exclusive
Catalog No. 50-352-918